Search
Search
| Gene Symbol: Mdga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199269.1 | NP_954890.1 | 12 - 13 | 2535 | 123 | |
| XM_008764688.1 | XP_008762910.1 | 12 - 13 | 3345 | 123 | ||
| GenBank mRNA | AY371924.1 | - | 12 - 13 | 2535 | 123 | |
Set Membership: |
|