Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01470236_g1
          See other MTR GE Assays ›
          Gene Symbol:
          Mtr
          Gene Name:
          5-methyltetrahydrofolate-homocysteine methyltransferase
          Gene Aliases:
          methioninesynthase
          Chromosome Location:
          Chr.17: 66210444 - 66295014 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01470236_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mtr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030864.1 NP_110491.1 32 - 33 3955 104
          GenBank mRNA AF034214.1 - 32 - 33 3955 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700002036.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mtr

          Gene Ontology Categories:

          Function(s) Process(es)

          protein methylation
          methionine metabolic process
          methionine biosynthetic process
          methylation
          pteridine-containing compound metabolic process
          tetrahydrofolate metabolic process
          homocysteine metabolic process
          cellular response to methionine
          folic acid binding
          methyltransferase activity
          zinc ion binding
          methionine synthase activity
          S-adenosylmethionine-homocysteine S-methyltransferase activity
          amino acid binding
          cobalamin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P479917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291454 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7808 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58552 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137090 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88548 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99822 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92954 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10392 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P479917 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P479917 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P479917 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P479917 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 392 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53394 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6635 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14118 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15286 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28144 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22439 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10795789 - -
          Affymetrix Rat Gene 1.0 ST Array 10795777 - -
          Affymetrix Rat Gene 1.0 ST Array 10795776 - -
          Affymetrix Rat Gene 1.0 ST Array 10795783 - -
          Affymetrix Rat Gene 1.0 ST Array 10795767 - -
          Affymetrix Rat Gene 1.0 ST Array 10795762 - -
          Affymetrix Rat Gene 1.0 ST Array 10795786 - -
          Affymetrix Rat Gene 1.0 ST Array 10795770 - -
          Affymetrix Rat Gene 1.0 ST Array 10795761 - -
          Affymetrix Rat Gene 1.0 ST Array 10795780 - -
          Affymetrix Rat Gene 1.0 ST Array 10795775 - -
          Affymetrix Rat Gene 1.0 ST Array 10795771 - -
          Affymetrix Rat Gene 1.0 ST Array 10795792 - -
          Affymetrix Rat Gene 1.0 ST Array 10795778 - -
          Affymetrix Rat Gene 1.0 ST Array 10795785 - -
          Affymetrix Rat Gene 1.0 ST Array 10795772 - -
          Affymetrix Rat Gene 1.0 ST Array 10795773 - -
          Affymetrix Rat Gene 1.0 ST Array 10795782 - -
          Affymetrix Rat Gene 1.0 ST Array 10795787 - -
          Affymetrix Rat Gene 1.0 ST Array 10795788 - -
          Affymetrix Rat Gene 1.0 ST Array 10795790 - -
          Affymetrix Rat Gene 1.0 ST Array 10795793 - -
          Affymetrix Rat Gene 1.0 ST Array 10795764 - -
          Affymetrix Rat Gene 1.0 ST Array 10795768 - -
          Affymetrix Rat Gene 1.0 ST Array 10795784 - -
          Affymetrix Rat Gene 1.0 ST Array 10795765 - -
          Affymetrix Rat Gene 1.0 ST Array 10795766 - -
          Affymetrix Rat Gene 1.0 ST Array 10795791 - -
          Affymetrix Rat Gene 1.0 ST Array 10795769 - -
          Affymetrix Rat Gene 1.0 ST Array 10795763 - -
          Affymetrix Rat Gene 1.0 ST Array 10795781 - -
          Affymetrix Rat Gene 1.0 ST Array 10795774 - -
          Affymetrix Rat Gene 1.0 ST Array 10795779 - -
          Affymetrix Rat Gene 1.1 ST Array 10795760 - -
          Rat Genome 230 2.0 Array 1387545_PM_at - -
          Rat Genome 230 2.0 Array 1387545_at - -
          Affymetrix HT Rat Focus Array 1387545_PM_at - -
          Affymetrix HT Rat Focus Array 1387545_at - -
          Affymetrix Rat Clariom S Assay PSR1700013889.rn.1 TC1700002036.rn.1 RTA TC
          Rat Expression Set 230 1387545_PM_at - -
          Rat Expression Set 230 1387545_at - -
          Affymetrix Rat Gene 1.0 ST Array 10795760 - -
          Rat Genome U34 Set AF034214_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21563506 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373194 - -
          RatRef-12 Expression BeadChip ILMN_69292 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline