Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01470643_m1
See other WNT3A GE Assays ›
Gene Symbol:
Wnt3a
Gene Name:
wingless-type MMTV integration site family, member 3A
Gene Aliases:
-
Chromosome Location:
Chr.10: 45598898 - 45643151 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01470643_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Wnt3a
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001107005.2
NP_001100475.2
3 - 4
598
76
XM_006246487.2
XP_006246549.1
3 - 4
756
76
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1000002906.rn.1
Set Membership:
Amplicon length between 71 and 85
3' Most
Recommended Assay
Rattus norvegicus
1700
Inventoried
Probe spans exons
Panther Classification:
Molecular Function -
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
in utero embryonic development
somitogenesis
positive regulation of cytokine production
positive regulation of protein phosphorylation
heart looping
positive regulation of receptor internalization
negative regulation of heart induction by canonical Wnt signaling pathway
determination of left/right symmetry
axonogenesis
axon guidance
mesoderm development
biological_process
positive regulation of cell proliferation
anterior/posterior pattern specification
COP9 signalosome assembly
negative regulation of neuron projection development
Wnt signaling pathway
spinal cord association neuron differentiation
hippocampus development
cell proliferation in forebrain
Wnt signaling pathway involved in forebrain neuroblast division
dorsal/ventral neural tube patterning
hemopoiesis
platelet activation
neuron differentiation
extracellular matrix organization
mammary gland development
positive regulation of B cell proliferation
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
cellular protein localization
positive regulation of catenin import into nucleus
skeletal muscle cell differentiation
post-anal tail morphogenesis
inner ear morphogenesis
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
cell fate commitment
regulation of cell differentiation
negative regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
somatic stem cell division
positive regulation of mesodermal cell fate specification
paraxial mesodermal cell fate commitment
positive regulation of collateral sprouting in absence of injury
negative regulation of axon extension involved in axon guidance
negative regulation of neurogenesis
regulation of axonogenesis
positive regulation of sequence-specific DNA binding transcription factor activity
palate development
canonical Wnt signaling pathway
cardiac muscle cell fate commitment
positive regulation of protein tyrosine kinase activity
positive regulation of dermatome development
canonical Wnt signaling pathway involved in cardiac muscle cell fate commitment
regulation of microtubule cytoskeleton organization
platelet aggregation
axis elongation involved in somitogenesis
positive regulation of canonical Wnt signaling pathway
positive regulation of cell-cell adhesion mediated by cadherin
positive regulation of canonical Wnt signaling pathway involved in controlling type B pancreatic cell proliferation
positive regulation of neural precursor cell proliferation
positive regulation of cardiac muscle cell differentiation
molecular_function
transcription coactivator activity
frizzled binding
frizzled-2 binding
protein domain specific binding
receptor agonist activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v2
A_64_P100051
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P100051
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P100051
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_64_P100051
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_44_P534762
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR1000025507.rn.1
TC1000002906.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1000025506.rn.1
TC1000002906.rn.1
RTA TC
Array Name
Probe IDs
Targets
Target Type
ABI Rat Genome Survey Microarray
20759651
-
-
Array Name
Probe IDs
Targets
Target Type
Illumina ratRef-12 v1.0 expression beadchip
ILMN_1362833
-
-
RatRef-12 Expression BeadChip
ILMN_50813
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline