Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01470881_m1
          See other SLU7 GE Assays ›
          Gene Symbol:
          Slu7
          Gene Name:
          SLU7 splicing factor homolog (S. cerevisiae)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 29029511 - 29044919 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01470881_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slu7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100550.1 NP_001094020.1 15 - 16 1656 126
          XM_008767640.1 XP_008765862.1 14 - 15 2431 126
          XM_008767641.1 XP_008765863.1 15 - 16 2103 126
          XM_008767642.1 XP_008765864.1 15 - 16 2308 126
          GenBank mRNA BC158865.1 - 15 - 16 1622 7938574 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000416.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA splicing, via transesterification reactions
          alternative mRNA splicing, via spliceosome
          mRNA 3'-splice site recognition
          intracellular protein transport
          RNA splicing
          cellular response to heat
          second spliceosomal transesterification activity
          zinc ion binding
          pre-mRNA 3'-splice site binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P117726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756631 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88820 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79902 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29524 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163243 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70232 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170987 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160874 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135290 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117726 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117726 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P117726 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18596 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17341 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66146 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21489 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43078 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10732877 - -
          Affymetrix Rat Gene 1.0 ST Array 10732882 - -
          Affymetrix Rat Gene 1.0 ST Array 10732881 - -
          Affymetrix Rat Gene 1.0 ST Array 10732874 - -
          Affymetrix Rat Gene 1.0 ST Array 10732875 - -
          Affymetrix Rat Gene 1.0 ST Array 10732884 - -
          Affymetrix Rat Gene 1.0 ST Array 10732873 - -
          Affymetrix Rat Gene 1.0 ST Array 10732871 - -
          Affymetrix Rat Gene 1.0 ST Array 10732880 - -
          Affymetrix Rat Gene 1.0 ST Array 10732886 - -
          Affymetrix Rat Gene 1.0 ST Array 10732887 - -
          Affymetrix Rat Gene 1.0 ST Array 10732883 - -
          Affymetrix Rat Gene 1.0 ST Array 10732878 - -
          Affymetrix Rat Gene 1.0 ST Array 10732879 - -
          Affymetrix Rat Gene 1.0 ST Array 10732876 - -
          Affymetrix Rat Gene 1.0 ST Array 10732888 - -
          Affymetrix Rat Gene 1.0 ST Array 10732885 - -
          Affymetrix Rat Gene 1.1 ST Array 10732869 - -
          Rat Genome 230 2.0 Array 1398588_PM_at - -
          Rat Genome 230 2.0 Array 1381030_PM_at - -
          Rat Genome 230 2.0 Array 1381030_at - -
          Rat Genome 230 2.0 Array 1398588_at - -
          Affymetrix HT Rat Focus Array 1374007_PM_at - -
          Affymetrix HT Rat Focus Array 1381030_at - -
          Affymetrix HT Rat Focus Array 1381030_PM_at - -
          Affymetrix HT Rat Focus Array 1374007_at - -
          Affymetrix HT Rat Focus Array 1398588_at - -
          Affymetrix HT Rat Focus Array 1398588_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000003578.rn.1 TC1000000416.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003579.rn.1 TC1000000416.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10732869 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355193 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline