Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01470916_g1
          See other TBC1D24 GE Assays ›
          Gene Symbol:
          Tbc1d24
          Gene Name:
          TBC1 domain family, member 24
          Gene Aliases:
          RGD1306143
          Chromosome Location:
          Chr.10: 13551100 - 13576739 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01470916_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tbc1d24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105769.1 NP_001099239.1 6 - 7 1568 83
          XM_006245926.1 XP_006245988.1 5 - 6 1327 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002382.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s)

          neuron projection development
          regulation of cilium assembly

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P523289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646760 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129527 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17854 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120264 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P523289 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P523289 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P523289 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P523289 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87653 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7081 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741006 - -
          Affymetrix Rat Gene 1.0 ST Array 10741002 - -
          Affymetrix Rat Gene 1.0 ST Array 10741008 - -
          Affymetrix Rat Gene 1.0 ST Array 10741005 - -
          Affymetrix Rat Gene 1.0 ST Array 10741007 - -
          Affymetrix Rat Gene 1.0 ST Array 10741003 - -
          Affymetrix Rat Gene 1.0 ST Array 10741004 - -
          Affymetrix Rat Gene 1.1 ST Array 10741001 - -
          Rat Genome 230 2.0 Array 1378146_PM_at - -
          Rat Genome 230 2.0 Array 1378146_at - -
          Affymetrix Rat Clariom S Assay PSR1000021041.rn.1 TC1000002382.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021040.rn.1 TC1000002382.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021049.rn.1 TC1000002382.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021039.rn.1 TC1000002382.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021043.rn.1 TC1000002382.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021044.rn.1 TC1000002382.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021047.rn.1 TC1000002382.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021042.rn.1 TC1000002382.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021048.rn.1 TC1000002382.rn.1 RTA TC
          Rat Expression Set 230 1378146_at - -
          Rat Expression Set 230 1378146_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741001 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20723196 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370531 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline