Search
Search

RGD1307929| Gene Symbol: RGD1307929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768148.1 | XP_008766370.1 | 27 - 28 | 5187 | 60 | |
| XM_220636.7 | XP_220636.3 | 27 - 28 | 5187 | 60 | ||
Set Membership: |
|