Search
Search
| Gene Symbol: Tnmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022290.1 | NP_071626.1 | 5 - 6 | 794 | 85 | |
| GenBank mRNA | AB055423.1 | - | 5 - 6 | 583 | 85 | |
| AF191769.1 | - | 5 - 6 | 794 | 85 | ||
Set Membership: |
|