Search
Search
| Gene Symbol: Nlrc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764491.1 | XP_008762713.1 | 5 - 6 | 2578 | 136 | |
| XM_008764492.1 | XP_008762714.1 | 5 - 6 | 2578 | 136 | ||
| XM_008764493.1 | XP_008762715.1 | 5 - 6 | 2578 | 136 | ||
Set Membership: |
|