Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01473755_m1
          See other SPAST GE Assays ›
          Gene Symbol:
          Spast
          Gene Name:
          spastin
          Gene Aliases:
          Spg4
          Chromosome Location:
          Chr.6: 22230067 - 22282164 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01473755_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spast
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108702.2 NP_001102172.2 15 - 16 1627 98
          XM_008764486.1 XP_008762708.1 16 - 17 2003 98
          XM_008764487.1 XP_008762709.1 16 - 17 2000 98
          XM_008764488.1 XP_008762710.1 15 - 16 1908 98
          GenBank mRNA BC166846.1 - 15 - 16 1627 9030077 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001964.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule bundle formation
          ER to Golgi vesicle-mediated transport
          axonogenesis
          metabolic process
          positive regulation of microtubule depolymerization
          cytoplasmic microtubule organization
          protein hexamerization
          microtubule severing
          protein homooligomerization
          ATP binding
          microtubule binding
          microtubule-severing ATPase activity
          alpha-tubulin binding
          beta-tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P820087 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P21124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156597 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48786 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129948 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174028 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21124 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P820087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P820087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21124 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P21124 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P820087 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21124 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11858 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10888588 - -
          Affymetrix Rat Gene 1.0 ST Array 10888592 - -
          Affymetrix Rat Gene 1.0 ST Array 10888585 - -
          Affymetrix Rat Gene 1.0 ST Array 10888593 - -
          Affymetrix Rat Gene 1.0 ST Array 10888590 - -
          Affymetrix Rat Gene 1.0 ST Array 10888594 - -
          Affymetrix Rat Gene 1.0 ST Array 10888586 - -
          Affymetrix Rat Gene 1.0 ST Array 10888589 - -
          Affymetrix Rat Gene 1.0 ST Array 10888583 - -
          Affymetrix Rat Gene 1.0 ST Array 10888587 - -
          Affymetrix Rat Gene 1.0 ST Array 10888584 - -
          Affymetrix Rat Gene 1.0 ST Array 10888581 - -
          Affymetrix Rat Gene 1.0 ST Array 10888591 - -
          Affymetrix Rat Gene 1.0 ST Array 10888595 - -
          Affymetrix Rat Gene 1.0 ST Array 10888582 - -
          Affymetrix Rat Gene 1.0 ST Array 10888580 - -
          Affymetrix Rat Gene 1.1 ST Array 10888579 - -
          Rat Genome 230 2.0 Array 1383782_at - -
          Rat Genome 230 2.0 Array 1383782_PM_at - -
          Rat Genome 230 2.0 Array 1390893_PM_at - -
          Rat Genome 230 2.0 Array 1390893_at - -
          Rat Genome 230 2.0 Array 1383192_at - -
          Rat Genome 230 2.0 Array 1383192_PM_at - -
          Affymetrix HT Rat Focus Array 1383782_PM_at - -
          Affymetrix HT Rat Focus Array 1383192_at - -
          Affymetrix HT Rat Focus Array 1383192_PM_at - -
          Affymetrix HT Rat Focus Array 1383782_at - -
          Affymetrix Rat Clariom S Assay PSR0600013899.rn.1 TC0600001964.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600013900.rn.1 TC0600001964.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600013898.rn.1 TC0600001964.rn.1 RTA TC
          Rat Expression Set 230 1383192_at - -
          Rat Expression Set 230 1383192_PM_at - -
          Rat Expression Set 230 1383782_PM_at - -
          Rat Expression Set 230 1383782_at - -
          Affymetrix Rat Gene 1.0 ST Array 10888579 - -
          Rat Genome U34 Set rc_AA875194_at - -
          Rat Genome U34 Set AA686361_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21749666 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351249 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline