Search
Search
| Gene Symbol: Sash3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134992.1 | NP_001128464.1 | 1 - 2 | 211 | 74 | |
| XM_006257530.2 | XP_006257592.1 | 1 - 2 | 265 | 74 | ||
| GenBank mRNA | BC169091.1 | - | 1 - 2 | 211 | 7933144 | 74 |
Set Membership: |
|