Search
Search
| Gene Symbol: Dll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053666.1 | NP_446118.1 | 5 - 6 | 895 | 91 | |
| XM_006228611.2 | XP_006228673.1 | 5 - 6 | 954 | 91 | ||
| GenBank mRNA | AF084576.1 | - | 5 - 6 | 895 | 91 | |
Set Membership: |
|