Search
Search

Izumo2
LOC100911976| Gene Symbol: Izumo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229184.2 | XP_006229246.1 | 3 - 4 | 404 | 190 | |
| XM_008759452.1 | XP_008757674.1 | 2 - 3 | 329 | 190 | ||
| Gene Symbol: LOC100911976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228963.2 | XP_006229025.1 | 3 - 4 | 404 | 190 | |
| XM_008759430.1 | XP_008757652.1 | 2 - 3 | 329 | 190 | ||
Set Membership: |
|