Search
Search
| Gene Symbol: Rtn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201562.3 | NP_963856.1 | 2 - 3 | 74 | 68 | |
| GenBank mRNA | AY279211.1 | - | 2 - 3 | 74 | 68 | |
| BC129079.1 | - | 2 - 3 | 189 | 8370402 | 68 | |
Set Membership: |
|