Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01476367_m1
See other APOE GE Assays ›
Gene Symbol:
Apoe
Gene Name:
apolipoprotein E
Gene Aliases:
APOEA
Chromosome Location:
Chr.1: 80612894 - 80616820 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01476367_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Apoe
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001270681.1
NP_001257610.1
1 - 2
115
61
GenBank mRNA
FQ220125.1
-
1 - 2
40
61
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001270681.1 transcripts.
Set Membership:
Amplicon length less than or equal to 70
Rattus norvegicus
Probe spans exons
Panther Classification:
Molecular Function -
apolipoprotein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of endothelial cell proliferation
response to dietary excess
lipid metabolic process
triglyceride metabolic process
cholesterol biosynthetic process
cholesterol catabolic process
lipid transport
cellular calcium ion homeostasis
receptor-mediated endocytosis
response to oxidative stress
G-protein coupled receptor signaling pathway
nitric oxide mediated signal transduction
aging
protein localization
cholesterol metabolic process
regulation of gene expression
negative regulation of platelet activation
positive regulation of cholesterol esterification
positive regulation of cholesterol efflux
peripheral nervous system axon regeneration
long-chain fatty acid transport
protein import
triglyceride catabolic process
cGMP-mediated signaling
negative regulation of blood coagulation
positive regulation of cGMP biosynthetic process
neuron projection regeneration
axon regeneration
regulation of cholesterol transport
regulation of Cdc42 protein signal transduction
response to retinoic acid
positive regulation of low-density lipoprotein particle receptor catabolic process
response to insulin
cholesterol efflux
phospholipid efflux
very-low-density lipoprotein particle remodeling
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
high-density lipoprotein particle assembly
chylomicron remnant clearance
high-density lipoprotein particle clearance
very-low-density lipoprotein particle clearance
lipoprotein metabolic process
lipoprotein biosynthetic process
lipoprotein catabolic process
vasodilation
cholesterol homeostasis
positive regulation of catalytic activity
negative regulation of MAP kinase activity
negative regulation of neuron apoptotic process
negative regulation of blood vessel endothelial cell migration
reverse cholesterol transport
response to ethanol
negative regulation of cholesterol biosynthetic process
positive regulation of axon extension
positive regulation of lipid biosynthetic process
oligodendrocyte differentiation
artery morphogenesis
negative regulation of inflammatory response
positive regulation of nitric-oxide synthase activity
positive regulation of membrane protein ectodomain proteolysis
negative regulation of lipid biosynthetic process
maintenance of location in cell
lipid homeostasis
fatty acid homeostasis
positive regulation of dendritic spine development
negative regulation of dendritic spine development
cellular response to interleukin-1
cellular response to growth factor stimulus
cellular response to cholesterol
cardiovascular system development
negative regulation of cholesterol efflux
AMPA glutamate receptor clustering
NMDA glutamate receptor clustering
regulation of beta-amyloid clearance
regulation of neuron death
negative regulation of neuron death
positive regulation of neuron death
negative regulation of postsynaptic membrane organization
positive regulation of postsynaptic membrane organization
negative regulation of presynaptic membrane organization
positive regulation of presynaptic membrane organization
positive regulation of beta-amyloid formation
negative regulation of beta-amyloid formation
regulation of tau-protein kinase activity
negative regulation of dendritic spine maintenance
positive regulation of dendritic spine maintenance
positive regulation of phospholipid efflux
positive regulation of neurofibrillary tangle assembly
negative regulation of phospholipid efflux
negative regulation of lipid transport across blood brain barrier
positive regulation of lipid transport across blood brain barrier
beta-amyloid binding
receptor binding
lipid transporter activity
phospholipid binding
heparin binding
lipid binding
cholesterol binding
antioxidant activity
cholesterol transporter activity
identical protein binding
protein homodimerization activity
hydroxyapatite binding
metal chelating activity
tau protein binding
low-density lipoprotein particle receptor binding
phosphatidylcholine-sterol O-acyltransferase activator activity
very-low-density lipoprotein particle receptor binding
lipoprotein particle binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR0100039029.rn.1
TC0100004935.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline