Search
Search
| Gene Symbol: Fuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037646.1 | NP_001032735.1 | 1 - 2 | 250 | 120 | |
| XM_006229059.2 | XP_006229121.1 | 1 - 2 | 289 | 120 | ||
| XM_006229060.2 | XP_006229122.1 | 1 - 2 | 289 | 120 | ||
| XM_006229061.2 | XP_006229123.1 | 1 - 2 | 290 | 120 | ||
| XM_008759373.1 | XP_008757595.1 | 1 - 2 | 290 | 120 | ||
| XM_008759374.1 | XP_008757596.1 | 1 - 2 | 290 | 120 | ||
| GenBank mRNA | BC107809.1 | - | 1 - 2 | 250 | 7453736 | 120 |
| FQ228777.1 | - | 1 - 2 | 235 | 120 | ||
Set Membership: |
|