Search
Search
| Gene Symbol: Snrpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271323.1 | NP_001258252.1 | 4 - 5 | 428 | 184 | |
| XM_006229323.2 | XP_006229385.1 | 4 - 5 | 587 | 184 | ||
| GenBank mRNA | FM044635.1 | - | 4 - 5 | 412 | 184 | |
| FQ232647.1 | - | 4 - 5 | 413 | 184 | ||
Set Membership: |
|