Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01478183_m1
          See other IQGAP1 GE Assays ›
          Gene Symbol:
          Iqgap1
          Gene Name:
          IQ motif containing GTPase activating protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 142525041 - 142615673 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01478183_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Iqgap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108489.1 NP_001101959.2 15 - 16 1875 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005887.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein modulator protein-binding activity modulator GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cytokine production
          epidermal growth factor receptor signaling pathway
          small GTPase mediated signal transduction
          fibroblast growth factor receptor signaling pathway
          negative regulation of dephosphorylation
          cellular response to platelet-derived growth factor stimulus
          cellular response to fibroblast growth factor stimulus
          positive regulation of protein kinase activity
          platelet-derived growth factor receptor signaling pathway
          cellular response to calcium ion
          cellular response to epidermal growth factor stimulus
          positive regulation of protein serine/threonine kinase activity
          neuron projection extension
          protein binding
          calmodulin binding
          phosphatidylinositol-3,4,5-trisphosphate binding
          Rho GTPase binding
          protein kinase binding
          protein phosphatase binding
          protein complex binding
          protein serine/threonine kinase activator activity
          Rac GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P424087 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P856007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313836 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45377 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93491 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83833 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P856007 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P424087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P856007 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P424087 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P856007 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P424087 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P157645 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P424087 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P856007 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45542 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49299 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10723163 - -
          Affymetrix Rat Gene 1.0 ST Array 10723188 - -
          Affymetrix Rat Gene 1.0 ST Array 10723164 - -
          Affymetrix Rat Gene 1.0 ST Array 10723173 - -
          Affymetrix Rat Gene 1.0 ST Array 10723176 - -
          Affymetrix Rat Gene 1.0 ST Array 10723192 - -
          Affymetrix Rat Gene 1.0 ST Array 10723187 - -
          Affymetrix Rat Gene 1.0 ST Array 10723167 - -
          Affymetrix Rat Gene 1.0 ST Array 10723194 - -
          Affymetrix Rat Gene 1.0 ST Array 10723161 - -
          Affymetrix Rat Gene 1.0 ST Array 10723183 - -
          Affymetrix Rat Gene 1.0 ST Array 10723189 - -
          Affymetrix Rat Gene 1.0 ST Array 10723166 - -
          Affymetrix Rat Gene 1.0 ST Array 10723179 - -
          Affymetrix Rat Gene 1.0 ST Array 10723185 - -
          Affymetrix Rat Gene 1.0 ST Array 10723162 - -
          Affymetrix Rat Gene 1.0 ST Array 10723172 - -
          Affymetrix Rat Gene 1.0 ST Array 10723157 - -
          Affymetrix Rat Gene 1.0 ST Array 10723165 - -
          Affymetrix Rat Gene 1.0 ST Array 10723184 - -
          Affymetrix Rat Gene 1.0 ST Array 10723171 - -
          Affymetrix Rat Gene 1.0 ST Array 10723169 - -
          Affymetrix Rat Gene 1.0 ST Array 10723177 - -
          Affymetrix Rat Gene 1.0 ST Array 10723158 - -
          Affymetrix Rat Gene 1.0 ST Array 10723181 - -
          Affymetrix Rat Gene 1.0 ST Array 10723182 - -
          Affymetrix Rat Gene 1.0 ST Array 10723174 - -
          Affymetrix Rat Gene 1.0 ST Array 10723168 - -
          Affymetrix Rat Gene 1.0 ST Array 10723193 - -
          Affymetrix Rat Gene 1.0 ST Array 10723190 - -
          Affymetrix Rat Gene 1.0 ST Array 10723178 - -
          Affymetrix Rat Gene 1.0 ST Array 10723186 - -
          Affymetrix Rat Gene 1.0 ST Array 10723160 - -
          Affymetrix Rat Gene 1.0 ST Array 10723191 - -
          Affymetrix Rat Gene 1.0 ST Array 10723159 - -
          Affymetrix Rat Gene 1.0 ST Array 10723175 - -
          Affymetrix Rat Gene 1.0 ST Array 10723170 - -
          Affymetrix Rat Gene 1.0 ST Array 10723180 - -
          Affymetrix Rat Gene 1.1 ST Array 10723156 - -
          Rat Genome 230 2.0 Array 1388762_PM_at - -
          Rat Genome 230 2.0 Array 1388762_at - -
          Rat Genome 230 2.0 Array 1388764_PM_at - -
          Rat Genome 230 2.0 Array 1388764_at - -
          Affymetrix HT Rat Focus Array 1388762_PM_at - -
          Affymetrix HT Rat Focus Array 1388764_at - -
          Affymetrix HT Rat Focus Array 1388762_at - -
          Affymetrix HT Rat Focus Array 1388764_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100046980.rn.1 TC0100005887.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10723156 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20833580 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373759 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline