Search
Search

RGD1564788| Gene Symbol: RGD1564788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253254.2 | XP_006253316.1 | 29 - 30 | 4267 | 69 | |
| XM_006253256.2 | XP_006253318.1 | 29 - 30 | 3696 | 69 | ||
| XM_008771311.1 | XP_008769533.1 | 27 - 28 | 4267 | 69 | ||
Set Membership: |
|