Search
Search
| Gene Symbol: Dak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039031.1 | NP_001034120.1 | 6 - 7 | 839 | 114 | |
| XM_006231056.2 | XP_006231118.1 | 6 - 7 | 725 | 114 | ||
| XM_006231057.2 | XP_006231119.2 | 6 - 7 | 1008 | 114 | ||
| GenBank mRNA | BC098925.1 | - | 6 - 7 | 839 | 7112371 | 114 |
Set Membership: |
|