Search
Search
| Gene Symbol: A1cf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133400.1 | NP_596891.1 | 4 - 5 | 471 | 68 | |
| XM_006231272.2 | XP_006231334.1 | 4 - 5 | 642 | 68 | ||
| GenBank mRNA | AF290984.1 | - | 4 - 5 | 471 | 68 | |
| AF442133.1 | - | 4 - 5 | 471 | 68 | ||
| AF442134.1 | - | 4 - 5 | 471 | 68 | ||
| AY028945.1 | - | 3 - 4 | 369 | 68 | ||
Set Membership: |
|