Search
Search

Stambpl1| Gene Symbol: Stambpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115048.1 | NP_001108520.1 | 4 - 5 | 611 | 62 | |
| XM_006231296.2 | XP_006231358.1 | 4 - 5 | 658 | 62 | ||
| XM_006231297.2 | XP_006231359.1 | 4 - 5 | 423 | 62 | ||
| XM_006231299.2 | XP_006231361.1 | 4 - 5 | 795 | 62 | ||
| XM_008760346.1 | XP_008758568.1 | 3 - 4 | 380 | 62 | ||
| XM_008760347.1 | XP_008758569.1 | 2 - 3 | 636 | 62 | ||
| XM_008760349.1 | XP_008758571.1 | 3 - 4 | 411 | 62 | ||
| GenBank mRNA | BC158624.1 | - | 4 - 5 | 611 | 7454746 | 62 |
Set Membership: |
|