Search
Search

Bcl2l12| Gene Symbol: Bcl2l12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108480.1 | NP_001101950.1 | 3 - 4 | 380 | 122 | |
| XM_006229097.2 | XP_006229159.1 | 2 - 3 | 353 | 122 | ||
Set Membership: |
|