Search
Search
| Gene Symbol: Cyp2f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019303.1 | NP_062176.1 | 6 - 7 | 976 | 75 | |
| XM_006228585.2 | XP_006228647.1 | 7 - 8 | 1084 | 75 | ||
| GenBank mRNA | AF017393.1 | - | 6 - 7 | 976 | 75 | |
| BC070939.1 | - | 7 - 8 | 1020 | 7099393 | 75 | |
Set Membership: |
|