Search
Search
| Gene Symbol: Rilpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191665.1 | NP_001178594.1 | 1 - 2 | 518 | 60 | |
| XM_006249294.1 | XP_006249356.1 | 1 - 2 | 782 | 60 | ||
| XM_006249295.1 | XP_006249357.1 | 1 - 2 | 782 | 60 | ||
| XM_006249296.2 | XP_006249358.1 | 1 - 2 | 782 | 60 | ||
| XM_006249297.2 | XP_006249359.1 | 1 - 2 | 782 | 60 | ||
Set Membership: |
|