Search
Search
| Gene Symbol: Vsig10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271336.1 | NP_001258265.1 | 3 - 4 | 440 | 63 | |
| XM_006249437.2 | XP_006249499.1 | 2 - 3 | 2764 | 63 | ||
| XM_006249438.2 | XP_006249500.1 | 2 - 3 | 2764 | 63 | ||
Set Membership: |
|