Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01482289_m1
          See other THSD1 GE Assays ›
          Gene Symbol:
          Thsd1
          Gene Name:
          thrombospondin, type I, domain containing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 74680621 - 74712122 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01482289_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thsd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108878.1 NP_001102348.1 4 - 5 1374 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001924.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          hematopoietic progenitor cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P288443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240142 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115347 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27251 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288443 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288443 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P288443 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288443 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792484 - -
          Affymetrix Rat Gene 1.0 ST Array 10792487 - -
          Affymetrix Rat Gene 1.0 ST Array 10792485 - -
          Affymetrix Rat Gene 1.0 ST Array 10792488 - -
          Affymetrix Rat Gene 1.0 ST Array 10792486 - -
          Affymetrix Rat Gene 1.1 ST Array 10792483 - -
          Affymetrix Rat Clariom S Assay PSR1600014248.rn.1 TC1600001924.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600014247.rn.1 TC1600001924.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10792483 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20701924 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361537 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline