Search
Search
| Gene Symbol: Erlin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106088.2 | NP_001099558.1 | 2 - 3 | 156 | 135 | |
| XM_008771323.1 | XP_008769545.1 | 2 - 3 | 409 | 135 | ||
| XM_008771324.1 | XP_008769546.1 | 2 - 3 | 351 | 135 | ||
| GenBank mRNA | BC168668.1 | - | 2 - 3 | 156 | 7934033 | 135 |
Set Membership: |
|