Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01482815_m1
          See other PRAGMIN GE Assays ›
          Gene Symbol:
          Pragmin
          Gene Name:
          pragma of Rnd2
          Gene Aliases:
          RGD1311793
          Chromosome Location:
          Chr.16: 59666794 - 59721246 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01482815_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pragmin
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107315.1 NP_001100785.1 2 - 3 555 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001746.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          protein kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P165616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24402 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158366 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10074 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8517 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P165616 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P165616 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P165616 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P384506 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78010 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10791993 - -
          Affymetrix Rat Gene 1.0 ST Array 10791992 - -
          Affymetrix Rat Gene 1.0 ST Array 10791991 - -
          Affymetrix Rat Gene 1.0 ST Array 10791990 - -
          Affymetrix Rat Gene 1.0 ST Array 10791994 - -
          Affymetrix Rat Gene 1.1 ST Array 10791989 - -
          Rat Genome 230 2.0 Array 1391121_at - -
          Rat Genome 230 2.0 Array 1391121_PM_at - -
          Affymetrix HT Rat Focus Array 1391121_at - -
          Affymetrix HT Rat Focus Array 1391121_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600012980.rn.1 TC1600001746.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600012978.rn.1 TC1600001746.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600012977.rn.1 TC1600001746.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600012979.rn.1 TC1600001746.rn.1 RTA TC
          Rat Expression Set 230 1391121_at - -
          Rat Expression Set 230 1391121_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10791989 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650690 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline