Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01482854_m1
          See other TPTE2 GE Assays ›
          Gene Symbol:
          Tpte2
          Gene Name:
          transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
          Gene Aliases:
          Tpte
          Chromosome Location:
          Chr.16: 74558529 - 74662194 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01482854_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tpte2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108877.1 NP_001102347.1 22 - 23 1829 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001922.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Protein Phosphatase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transmembrane transport
          peptidyl-tyrosine dephosphorylation
          protein tyrosine phosphatase activity
          ion channel activity
          protein tyrosine/serine/threonine phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P121586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518384 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76710 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118793 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40070 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55866 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126796 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31676 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117955 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88883 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P121586 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P121586 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P121586 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P121586 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101189 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72009 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792463 - -
          Affymetrix Rat Gene 1.0 ST Array 10792479 - -
          Affymetrix Rat Gene 1.0 ST Array 10792477 - -
          Affymetrix Rat Gene 1.0 ST Array 10792468 - -
          Affymetrix Rat Gene 1.0 ST Array 10792478 - -
          Affymetrix Rat Gene 1.0 ST Array 10792457 - -
          Affymetrix Rat Gene 1.0 ST Array 10792473 - -
          Affymetrix Rat Gene 1.0 ST Array 10792464 - -
          Affymetrix Rat Gene 1.0 ST Array 10792470 - -
          Affymetrix Rat Gene 1.0 ST Array 10792474 - -
          Affymetrix Rat Gene 1.0 ST Array 10792480 - -
          Affymetrix Rat Gene 1.0 ST Array 10792482 - -
          Affymetrix Rat Gene 1.0 ST Array 10792476 - -
          Affymetrix Rat Gene 1.0 ST Array 10792472 - -
          Affymetrix Rat Gene 1.0 ST Array 10792460 - -
          Affymetrix Rat Gene 1.0 ST Array 10792459 - -
          Affymetrix Rat Gene 1.0 ST Array 10792469 - -
          Affymetrix Rat Gene 1.0 ST Array 10792475 - -
          Affymetrix Rat Gene 1.0 ST Array 10792462 - -
          Affymetrix Rat Gene 1.0 ST Array 10792471 - -
          Affymetrix Rat Gene 1.0 ST Array 10792466 - -
          Affymetrix Rat Gene 1.0 ST Array 10792465 - -
          Affymetrix Rat Gene 1.0 ST Array 10792458 - -
          Affymetrix Rat Gene 1.0 ST Array 10792481 - -
          Affymetrix Rat Gene 1.0 ST Array 10792467 - -
          Affymetrix Rat Gene 1.0 ST Array 10792461 - -
          Affymetrix Rat Gene 1.1 ST Array 10792456 - -
          Affymetrix Rat Clariom S Assay PSR1600014219.rn.1 TC1600001922.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10792456 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21457399 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372147 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline