Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01482858_m1
          See other TFDP1 GE Assays ›
          Gene Symbol:
          Tfdp1
          Gene Name:
          transcription factor Dp-1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 81088640 - 81127244 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01482858_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tfdp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025718.1 NP_001020889.1 3 - 4 283 62
          XM_006253451.2 XP_006253513.1 4 - 5 548 62
          XM_008771397.1 XP_008769619.1 5 - 6 551 62
          XM_008771398.1 XP_008769620.1 5 - 6 630 62
          XM_008771399.1 XP_008769621.1 5 - 6 458 62
          GenBank mRNA BC098789.1 - 3 - 4 283 7375514 62

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC098789.1, NM_001025718.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tfdp1

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          epidermis development
          anoikis
          positive regulation of transcription, DNA-templated
          regulation of DNA biosynthetic process
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          transcription factor binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P247139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66936 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111940 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87861 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P247139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P247139 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P247139 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P247139 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P249606 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95627 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44278 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73538 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40512 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32326 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792757 - -
          Affymetrix Rat Gene 1.0 ST Array 10792755 - -
          Affymetrix Rat Gene 1.0 ST Array 10792753 - -
          Affymetrix Rat Gene 1.0 ST Array 10792761 - -
          Affymetrix Rat Gene 1.0 ST Array 10792752 - -
          Affymetrix Rat Gene 1.0 ST Array 10792754 - -
          Affymetrix Rat Gene 1.0 ST Array 10792758 - -
          Affymetrix Rat Gene 1.0 ST Array 10792756 - -
          Affymetrix Rat Gene 1.0 ST Array 10792759 - -
          Affymetrix Rat Gene 1.1 ST Array 10792751 - -
          Rat Genome 230 2.0 Array 1388867_PM_at - -
          Rat Genome 230 2.0 Array 1388867_at - -
          Affymetrix HT Rat Focus Array 1388867_at - -
          Affymetrix HT Rat Focus Array 1388867_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600007243.rn.1 TC1600000964.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600007244.rn.1 TC1600000964.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600007245.rn.1 TC1600000964.rn.1 RTA TC
          Rat Expression Set 230 1388867_PM_at - -
          Rat Expression Set 230 1388867_at - -
          Affymetrix Rat Gene 1.0 ST Array 10792751 - -
          Rat Genome U34 Set rc_AI227852_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21084743 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360118 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline