Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01482928_m1
          See other COL4A1 GE Assays ›
          Gene Symbol:
          Col4a1
          Gene Name:
          collagen, type IV, alpha 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 83522162 - 83632153 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01482928_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Col4a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135009.1 NP_001128481.1 51 - 52 5131 136
          GenBank mRNA BC089096.1 - 5 - 6 712 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001009.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          patterning of blood vessels
          brain development
          neuromuscular junction development
          epithelial cell differentiation
          blood vessel morphogenesis
          retinal blood vessel morphogenesis
          renal tubule morphogenesis
          cellular response to amino acid stimulus
          basement membrane organization
          molecular_function
          extracellular matrix structural constituent
          platelet-derived growth factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P149501 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P293606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378307 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135025 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22962 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92686 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116753 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56686 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P149501 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P293606 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P149501 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P293606 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P293606 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P149501 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1021644 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1070510 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 125 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10789597 - -
          Affymetrix Rat Gene 1.0 ST Array 10789647 - -
          Affymetrix Rat Gene 1.0 ST Array 10789644 - -
          Affymetrix Rat Gene 1.0 ST Array 10789609 - -
          Affymetrix Rat Gene 1.0 ST Array 10789612 - -
          Affymetrix Rat Gene 1.0 ST Array 10789613 - -
          Affymetrix Rat Gene 1.0 ST Array 10789632 - -
          Affymetrix Rat Gene 1.0 ST Array 10789610 - -
          Affymetrix Rat Gene 1.0 ST Array 10789646 - -
          Affymetrix Rat Gene 1.0 ST Array 10789600 - -
          Affymetrix Rat Gene 1.0 ST Array 10789608 - -
          Affymetrix Rat Gene 1.0 ST Array 10789604 - -
          Affymetrix Rat Gene 1.0 ST Array 10789627 - -
          Affymetrix Rat Gene 1.0 ST Array 10789636 - -
          Affymetrix Rat Gene 1.0 ST Array 10789652 - -
          Affymetrix Rat Gene 1.0 ST Array 10789639 - -
          Affymetrix Rat Gene 1.0 ST Array 10789629 - -
          Affymetrix Rat Gene 1.0 ST Array 10789633 - -
          Affymetrix Rat Gene 1.0 ST Array 10789621 - -
          Affymetrix Rat Gene 1.0 ST Array 10789595 - -
          Affymetrix Rat Gene 1.0 ST Array 10789626 - -
          Affymetrix Rat Gene 1.0 ST Array 10789603 - -
          Affymetrix Rat Gene 1.0 ST Array 10789630 - -
          Affymetrix Rat Gene 1.0 ST Array 10789637 - -
          Affymetrix Rat Gene 1.0 ST Array 10789611 - -
          Affymetrix Rat Gene 1.0 ST Array 10789645 - -
          Affymetrix Rat Gene 1.0 ST Array 10789634 - -
          Affymetrix Rat Gene 1.0 ST Array 10789641 - -
          Affymetrix Rat Gene 1.0 ST Array 10789618 - -
          Affymetrix Rat Gene 1.0 ST Array 10789640 - -
          Affymetrix Rat Gene 1.0 ST Array 10789596 - -
          Affymetrix Rat Gene 1.0 ST Array 10789599 - -
          Affymetrix Rat Gene 1.0 ST Array 10789651 - -
          Affymetrix Rat Gene 1.0 ST Array 10789649 - -
          Affymetrix Rat Gene 1.0 ST Array 10789620 - -
          Affymetrix Rat Gene 1.0 ST Array 10789628 - -
          Affymetrix Rat Gene 1.0 ST Array 10789624 - -
          Affymetrix Rat Gene 1.0 ST Array 10789625 - -
          Affymetrix Rat Gene 1.0 ST Array 10789602 - -
          Affymetrix Rat Gene 1.0 ST Array 10789623 - -
          Affymetrix Rat Gene 1.0 ST Array 10789631 - -
          Affymetrix Rat Gene 1.0 ST Array 10789606 - -
          Affymetrix Rat Gene 1.0 ST Array 10789619 - -
          Affymetrix Rat Gene 1.0 ST Array 10789635 - -
          Affymetrix Rat Gene 1.0 ST Array 10789643 - -
          Affymetrix Rat Gene 1.0 ST Array 10789601 - -
          Affymetrix Rat Gene 1.0 ST Array 10789605 - -
          Affymetrix Rat Gene 1.0 ST Array 10789598 - -
          Affymetrix Rat Gene 1.0 ST Array 10789622 - -
          Affymetrix Rat Gene 1.0 ST Array 10789638 - -
          Affymetrix Rat Gene 1.0 ST Array 10789650 - -
          Affymetrix Rat Gene 1.0 ST Array 10789642 - -
          Affymetrix Rat Gene 1.0 ST Array 10789607 - -
          Affymetrix Rat Gene 1.0 ST Array 10789616 - -
          Affymetrix Rat Gene 1.0 ST Array 10789615 - -
          Affymetrix Rat Gene 1.0 ST Array 10789617 - -
          Affymetrix Rat Gene 1.0 ST Array 10789614 - -
          Affymetrix Rat Gene 1.0 ST Array 10789648 - -
          Affymetrix Rat Gene 1.1 ST Array 10789594 - -
          Rat Genome 230 2.0 Array 1372439_PM_at - -
          Rat Genome 230 2.0 Array 1372439_at - -
          Rat Genome 230 2.0 Array 1373245_PM_at - -
          Rat Genome 230 2.0 Array 1373245_at - -
          Affymetrix HT Rat Focus Array 1372439_PM_at - -
          Affymetrix HT Rat Focus Array 1373245_at - -
          Affymetrix HT Rat Focus Array 1372439_at - -
          Affymetrix HT Rat Focus Array 1373245_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600007679.rn.1 TC1600001009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600007678.rn.1 TC1600001009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600007680.rn.1 TC1600001009.rn.1 RTA TC
          Rat Expression Set 230 1373245_at - -
          Rat Expression Set 230 1372439_at - -
          Rat Expression Set 230 1373245_PM_at - -
          Rat Expression Set 230 1372439_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10789594 - -
          Rat Genome U34 Set rc_AA964892_at - -
          Rat Genome U34 Set rc_AI176393_at - -
          Rat Genome U34 Set rc_AI175875_s_at - -
          Rat Genome U34 Set rc_AI175875_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22117229 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline