Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01482944_m1
          See other LOC290876 GE Assays ›
          Gene Symbol:
          LOC290876
          Gene Name:
          similar to RIKEN cDNA 1700029H14
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 81301810 - 81314707 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01482944_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC290876
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037182.1 NP_001032259.1 8 - 9 926 61
          XM_006253430.2 XP_006253492.1 8 - 9 1135 61
          XM_006253433.2 XP_006253495.1 8 - 9 930 61
          XM_008771386.1 XP_008769608.1 8 - 9 1174 61
          GenBank mRNA AB714639.1 - 2 - 3 159 61
          BC085810.1 - 8 - 9 886 61
          BC100249.1 - 8 - 9 926 7135908 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600002010.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC290876

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P22695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646372 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122278 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22695 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22695 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P22695 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18871 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15152 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10789435 - -
          Affymetrix Rat Gene 1.0 ST Array 10789434 - -
          Affymetrix Rat Gene 1.0 ST Array 10789440 - -
          Affymetrix Rat Gene 1.0 ST Array 10789438 - -
          Affymetrix Rat Gene 1.0 ST Array 10789437 - -
          Affymetrix Rat Gene 1.0 ST Array 10789433 - -
          Affymetrix Rat Gene 1.0 ST Array 10789441 - -
          Affymetrix Rat Gene 1.0 ST Array 10789432 - -
          Affymetrix Rat Gene 1.0 ST Array 10789439 - -
          Affymetrix Rat Gene 1.1 ST Array 10789431 - -
          Rat Genome 230 2.0 Array 1385348_PM_at - -
          Rat Genome 230 2.0 Array 1385348_at - -
          Affymetrix HT Rat Focus Array 1385348_at - -
          Affymetrix HT Rat Focus Array 1385348_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600014838.rn.1 TC1600002010.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600014839.rn.1 TC1600002010.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600014841.rn.1 TC1600002010.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600014840.rn.1 TC1600002010.rn.1 RTA TC
          Rat Expression Set 230 1385348_PM_at - -
          Rat Expression Set 230 1385348_at - -
          Affymetrix Rat Gene 1.0 ST Array 10789431 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21084893 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364687 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline