Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01484035_m1
          See other FMO4 GE Assays ›
          Gene Symbol:
          Fmo4
          Gene Name:
          flavin containing monooxygenase 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 80685385 - 80703575 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01484035_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fmo4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144561.2 NP_653147.1 9 - 10 1562 126
          NM_144562.1 NP_653148.1 7 - 8 1167 126
          XM_006250144.2 XP_006250206.1 9 - 10 1497 126
          GenBank mRNA AF458416.1 - 9 - 10 1563 126
          AF458417.1 - 7 - 8 1167 126
          BC081721.1 - 9 - 10 1552 7107835 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001774.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fmo4

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          drug catabolic process
          oxidation-reduction process
          monooxygenase activity
          N,N-dimethylaniline monooxygenase activity
          flavin adenine dinucleotide binding
          NADP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P170368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813947 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25989 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P170368 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P170368 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P170368 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P170368 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28657 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87285 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769360 - -
          Affymetrix Rat Gene 1.0 ST Array 10769351 - -
          Affymetrix Rat Gene 1.0 ST Array 10769356 - -
          Affymetrix Rat Gene 1.0 ST Array 10769353 - -
          Affymetrix Rat Gene 1.0 ST Array 10769354 - -
          Affymetrix Rat Gene 1.0 ST Array 10769355 - -
          Affymetrix Rat Gene 1.0 ST Array 10769357 - -
          Affymetrix Rat Gene 1.0 ST Array 10769359 - -
          Affymetrix Rat Gene 1.0 ST Array 10769358 - -
          Affymetrix Rat Gene 1.0 ST Array 10769350 - -
          Affymetrix Rat Gene 1.0 ST Array 10769352 - -
          Affymetrix Rat Gene 1.1 ST Array 10769349 - -
          Rat Genome 230 2.0 Array 1395026_at - -
          Rat Genome 230 2.0 Array 1379885_at - -
          Rat Genome 230 2.0 Array 1379885_PM_at - -
          Rat Genome 230 2.0 Array 1395026_PM_at - -
          Affymetrix HT Rat Focus Array 1379885_PM_at - -
          Affymetrix HT Rat Focus Array 1395026_at - -
          Affymetrix HT Rat Focus Array 1379885_at - -
          Affymetrix HT Rat Focus Array 1395026_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300012554.rn.1 TC1300001774.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012553.rn.1 TC1300001774.rn.1 RTA TC
          Rat Expression Set 230 1395026_PM_at - -
          Rat Expression Set 230 1379885_PM_at - -
          Rat Expression Set 230 1395026_at - -
          Rat Expression Set 230 1379885_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769349 - -
          Rat Genome U34 Set rc_AI232461_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22320480 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375784 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376656 - -
          RatRef-12 Expression BeadChip ILMN_51217 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline