Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01484924_m1
See other BDNF GE Assays ›
Gene Symbol:
Bdnf
Gene Name:
brain-derived neurotrophic factor
Gene Aliases:
-
Chromosome Location:
Chr.3: 100768637 - 100819216 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01484924_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Inventoried
Catalog #
4331182
Price
2,694.00
Your Price
Online offer:
2694.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Bdnf
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_012513.4
NP_036645.2
1 - 2
639
106
GenBank mRNA
EF125675.1
-
1 - 2
639
106
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0300001371.rn.1
Additional Information:
This assay is in the 5'UTR region of the EF125675.1,NM_012513.4 transcripts.
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Apoptosis
Rattus norvegicus
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Bdnf
Panther Classification:
Molecular Function -
neurotrophic factor
Gene Ontology Categories:
Function(s)
Process(es)
ureteric bud development
behavioral fear response
response to hypoxia
chronic inflammatory response
mitochondrial electron transport, NADH to ubiquinone
transmembrane receptor protein tyrosine kinase signaling pathway
gamma-aminobutyric acid signaling pathway
nervous system development
negative regulation of neuroblast proliferation
axon guidance
axon target recognition
learning or memory
learning
circadian rhythm
feeding behavior
neuron recognition
response to light stimulus
response to light intensity
response to hormone
response to auditory stimulus
glutamate secretion
response to fluoxetine
response to activity
dendrite development
regulation of metabolic process
nerve development
neuron differentiation
regulation of axon extension
response to nutrient levels
response to food
negative regulation of synaptic transmission, GABAergic
positive regulation of peptidyl-serine phosphorylation
response to vitamin A
response to tumor necrosis factor
response to potassium ion
mechanoreceptor differentiation
response to drug
fear response
negative regulation of apoptotic process
regulation of neuron apoptotic process
negative regulation of neuron apoptotic process
positive regulation of neuron differentiation
negative regulation of striated muscle tissue development
regulation of retinal cell programmed cell death
regulation of synaptic plasticity
regulation of long-term neuronal synaptic plasticity
positive regulation of long-term neuronal synaptic plasticity
regulation of short-term neuronal synaptic plasticity
axon extension
inner ear development
cognition
positive regulation of sequence-specific DNA binding transcription factor activity
response to electrical stimulus
positive regulation of synapse assembly
response to hyperoxia
excitatory postsynaptic potential
inhibitory postsynaptic potential
negative regulation of cell death
taste bud development
cellular response to tumor necrosis factor
cellular response to norepinephrine stimulus
response to anesthetic
dendrite extension
negative regulation of neuron death
response to L-glutamate
response to nerve growth factor
cellular response to nerve growth factor stimulus
neuron projection extension
positive regulation of glucocorticoid receptor signaling pathway
neurotrophin TRKB receptor binding
growth factor activity
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR0300009218.rn.1
TC0300001371.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline