Search
Search
| Gene Symbol: Atf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031018.1 | NP_112280.1 | 11 - 12 | 1262 | 57 | |
| GenBank mRNA | AY724488.1 | - | 840 | 57 | ||
| M65148.1 | - | 6 - 7 | 657 | 57 | ||
| U38938.1 | - | 11 - 12 | 1262 | 57 | ||
Set Membership: |
|