Search
Search
| Gene Symbol: Ttbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107766.1 | NP_001101236.1 | 10 - 11 | 1278 | 114 | |
| XM_006234833.2 | XP_006234895.1 | 12 - 13 | 1711 | 114 | ||
| XM_006234834.2 | XP_006234896.1 | 12 - 13 | 1354 | 114 | ||
| XM_006234835.2 | XP_006234897.1 | 11 - 12 | 1637 | 114 | ||
| XM_006234836.2 | XP_006234898.1 | 10 - 11 | 1489 | 114 | ||
| XM_006234837.2 | XP_006234899.1 | 12 - 13 | 1357 | 114 | ||
Set Membership: |
|