Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01485789_m1
          See other TTBK2 GE Assays ›
          Gene Symbol:
          Ttbk2
          Gene Name:
          tau tubulin kinase 2
          Gene Aliases:
          Ttbk1
          Chromosome Location:
          Chr.3: 112677932 - 112789613 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01485789_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ttbk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107766.1 NP_001101236.1 13 - 14 3355 96
          XM_006234833.2 XP_006234895.1 15 - 16 3788 96
          XM_006234834.2 XP_006234896.1 15 - 16 3431 96
          XM_006234835.2 XP_006234897.1 14 - 15 3714 96
          XM_006234836.2 XP_006234898.1 13 - 14 3566 96
          XM_006234837.2 XP_006234899.1 15 - 16 3434 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004013.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Protein Kinase 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          smoothened signaling pathway
          regulation of cell shape
          peptidyl-serine phosphorylation
          cilium assembly
          protein serine/threonine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P994116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143892 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53443 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153164 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134880 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40742 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27364 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55678 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58747 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P994116 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P994116 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P994116 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P994116 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77681 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49639 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10848897 - -
          Affymetrix Rat Gene 1.0 ST Array 10848892 - -
          Affymetrix Rat Gene 1.0 ST Array 10848900 - -
          Affymetrix Rat Gene 1.0 ST Array 10848903 - -
          Affymetrix Rat Gene 1.0 ST Array 10848895 - -
          Affymetrix Rat Gene 1.0 ST Array 10848902 - -
          Affymetrix Rat Gene 1.0 ST Array 10848893 - -
          Affymetrix Rat Gene 1.0 ST Array 10848896 - -
          Affymetrix Rat Gene 1.0 ST Array 10848904 - -
          Affymetrix Rat Gene 1.0 ST Array 10848898 - -
          Affymetrix Rat Gene 1.0 ST Array 10848894 - -
          Affymetrix Rat Gene 1.0 ST Array 10848905 - -
          Affymetrix Rat Gene 1.0 ST Array 10848901 - -
          Affymetrix Rat Gene 1.1 ST Array 10848891 - -
          Rat Genome 230 2.0 Array 1384908_at - -
          Rat Genome 230 2.0 Array 1384908_PM_at - -
          Affymetrix HT Rat Focus Array 1384908_PM_at - -
          Affymetrix HT Rat Focus Array 1384908_at - -
          Affymetrix Rat Clariom S Assay PSR0300030694.rn.1 TC0300004013.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300030691.rn.1 TC0300004013.rn.1 RTA TC
          Rat Expression Set 230 1384908_PM_at - -
          Rat Expression Set 230 1384908_at - -
          Affymetrix Rat Gene 1.0 ST Array 10848891 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21188675 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363629 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline