Search
Search

Catsper2| Gene Symbol: Catsper2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012220.1 | NP_001012220.1 | 8 - 9 | 1366 | 87 | |
| XM_006234860.2 | XP_006234922.1 | 8 - 9 | 1292 | 87 | ||
| XM_008762187.1 | XP_008760409.1 | 8 - 9 | 1048 | 87 | ||
| GenBank mRNA | BC079422.1 | - | 8 - 9 | 1366 | 7114001 | 87 |
Set Membership: |
|