Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01487518_g1
          See other AKR1C2 GE Assays ›
          Gene Symbol:
          Akr1c2
          Gene Name:
          aldo-keto reductase family 1, member C2
          Gene Aliases:
          Akr1c21
          Chromosome Location:
          Chr.17: 69388337 - 69435160 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01487518_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akr1c2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013057.1 NP_001013075.1 7 - 8 876 133
          XM_006254183.2 XP_006254245.1 7 - 8 888 133
          XM_006254184.2 XP_006254246.1 7 - 8 867 133
          XM_006254185.2 XP_006254247.1 6 - 7 810 133
          GenBank mRNA BC078957.1 - 7 - 8 876 7110806 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700002053.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Akr1c2

          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin metabolic process
          G-protein coupled receptor signaling pathway
          digestion
          steroid metabolic process
          positive regulation of cell proliferation
          epithelial cell differentiation
          progesterone metabolic process
          daunorubicin metabolic process
          doxorubicin metabolic process
          positive regulation of protein kinase B signaling
          oxidation-reduction process
          cellular response to jasmonic acid stimulus
          cellular response to prostaglandin D stimulus
          alditol:NADP+ 1-oxidoreductase activity
          oxidoreductase activity
          oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
          phenanthrene 9,10-monooxygenase activity
          carboxylic acid binding
          bile acid binding
          steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          androsterone dehydrogenase activity
          ketosteroid monooxygenase activity
          trans-1,2-dihydrobenzene-1,2-diol dehydrogenase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P323773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234968 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107569 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15298 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16543 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P323773 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P323773 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P323773 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P323773 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36169 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42774 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32187 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10799318 - -
          Affymetrix Rat Gene 1.0 ST Array 10799323 - -
          Affymetrix Rat Gene 1.0 ST Array 10799321 - -
          Affymetrix Rat Gene 1.0 ST Array 10799320 - -
          Affymetrix Rat Gene 1.0 ST Array 10799319 - -
          Affymetrix Rat Gene 1.0 ST Array 10799325 - -
          Affymetrix Rat Gene 1.0 ST Array 10799324 - -
          Affymetrix Rat Gene 1.0 ST Array 10799327 - -
          Affymetrix Rat Gene 1.0 ST Array 10799322 - -
          Affymetrix Rat Gene 1.0 ST Array 10799326 - -
          Affymetrix Rat Gene 1.1 ST Array 10799317 - -
          Affymetrix Rat Clariom S Assay PSR1700014093.rn.1 TC1700002053.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700014099.rn.1 TC1700002053.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700014095.rn.1 TC1700002053.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700014097.rn.1 TC1700002053.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700014100.rn.1 TC1700002053.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10799317 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349692 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline