Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01490500_m1
          See other SLC19A2 GE Assays ›
          Gene Symbol:
          Slc19a2
          Gene Name:
          solute carrier family 19 (thiamine transporter), member 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 82552586 - 82566586 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01490500_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc19a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001030024.1 NP_001025195.1 3 - 4 1197 86
          GenBank mRNA BC099811.1 - 3 - 4 1197 7929969 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000653.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc19a2

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          drug transmembrane transport
          biological_process
          thiamine transport
          thiamine transmembrane transport
          molecular_function
          thiamine transmembrane transporter activity
          thiamine uptake transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P789739 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P074082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402213 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125335 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167422 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P789739 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074082 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074082 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P789739 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P074082 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P789739 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P789739 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18040 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2479 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5283 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74158 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10765254 - -
          Affymetrix Rat Gene 1.0 ST Array 10765257 - -
          Affymetrix Rat Gene 1.0 ST Array 10765255 - -
          Affymetrix Rat Gene 1.0 ST Array 10765249 - -
          Affymetrix Rat Gene 1.0 ST Array 10765250 - -
          Affymetrix Rat Gene 1.0 ST Array 10765252 - -
          Affymetrix Rat Gene 1.0 ST Array 10765251 - -
          Affymetrix Rat Gene 1.0 ST Array 10765256 - -
          Affymetrix Rat Gene 1.0 ST Array 10765253 - -
          Affymetrix Rat Gene 1.1 ST Array 10765248 - -
          Rat Genome 230 2.0 Array 1390863_at - -
          Rat Genome 230 2.0 Array 1390863_PM_at - -
          Affymetrix HT Rat Focus Array 1390863_at - -
          Affymetrix HT Rat Focus Array 1390863_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300004581.rn.1 TC1300000653.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004582.rn.1 TC1300000653.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004580.rn.1 TC1300000653.rn.1 RTA TC
          Rat Expression Set 230 1390863_PM_at - -
          Rat Expression Set 230 1390863_at - -
          Affymetrix Rat Gene 1.0 ST Array 10765248 - -
          Rat Genome U34 Set rc_AI233639_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20904032 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365726 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline