Search
Search
| Gene Symbol: Ndst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024361.1 | NP_077337.1 | 13 - 14 | 2868 | 63 | |
| XM_008772130.1 | XP_008770352.1 | 9 - 10 | 1256 | 63 | ||
| GenBank mRNA | M92042.1 | - | 13 - 14 | 2868 | 63 | |
Set Membership: |
|