Search
Search
| Gene Symbol: Cd74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013069.2 | NP_037201.1 | 6 - 7 | 673 | 67 | |
| GenBank mRNA | BC059152.1 | - | 6 - 7 | 654 | 6890709 | 67 |
| FQ219430.1 | - | 6 - 7 | 656 | 67 | ||
| FQ225098.1 | - | 6 - 7 | 648 | 67 | ||
| FQ225666.1 | - | 4 - 5 | 292 | 67 | ||
| FQ227357.1 | - | 6 - 7 | 650 | 67 | ||
| FQ227561.1 | - | 6 - 7 | 657 | 67 | ||
| FQ228116.1 | - | 6 - 7 | 656 | 67 | ||
| FQ231756.1 | - | 6 - 7 | 657 | 67 | ||
| FQ232412.1 | - | 6 - 7 | 698 | 67 | ||
| FQ232423.1 | - | 6 - 7 | 687 | 67 | ||
| FQ232867.1 | - | 6 - 7 | 657 | 67 | ||
| FQ232996.1 | - | 6 - 7 | 650 | 67 | ||
| FQ234457.1 | - | 6 - 7 | 659 | 67 | ||
| X13044.1 | - | 6 - 7 | 629 | 67 | ||
Set Membership: |
|