Search
Search
| Gene Symbol: Smad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019275.2 | NP_062148.1 | 7 - 8 | 1017 | 85 | |
| GenBank mRNA | AB010954.1 | - | 7 - 8 | 1017 | 85 | |
| AF056002.1 | - | 1089 | 85 | |||
Set Membership: |
|