Search
Search
| Gene Symbol: Prdm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772189.1 | XP_008770411.1 | 5 - 6 | 1233 | 68 | |
| XM_008772190.1 | XP_008770412.1 | 4 - 5 | 1507 | 68 | ||
| XM_008772191.1 | XP_008770413.1 | 5 - 6 | 1233 | 68 | ||
Set Membership: |
|