Search
Search

Tnfrsf1a| Gene Symbol: Tnfrsf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013091.1 | NP_037223.1 | 8 - 9 | 1008 | 99 | |
| XM_006237334.2 | XP_006237396.1 | 7 - 8 | 1243 | 99 | ||
| GenBank mRNA | AF329976.1 | - | 8 - 9 | 1007 | 99 | |
| AF329977.1 | - | 8 - 9 | 1007 | 99 | ||
| AF329978.1 | - | 8 - 9 | 1007 | 99 | ||
| AF329979.1 | - | 8 - 9 | 1007 | 99 | ||
| AF329980.1 | - | 8 - 9 | 1007 | 99 | ||
| AF329981.1 | - | 8 - 9 | 1007 | 99 | ||
| BC086413.1 | - | 8 - 9 | 1028 | 7110094 | 99 | |
| M63122.1 | - | 8 - 9 | 1008 | 99 | ||
Set Membership: |
|