Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01492369_m1
          See other BCL2L13 GE Assays ›
          Gene Symbol:
          Bcl2l13
          Gene Name:
          BCL2-like 13 (apoptosis facilitator)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 153385200 - 153438096 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01492369_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bcl2l13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107885.1 NP_001101355.1 2 - 3 310 81
          XM_006237248.1 XP_006237310.1 2 - 3 314 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001695.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P18010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94971 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149468 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37261 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29061 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91557 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20523 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18010 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18010 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P18010 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17344 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4476 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10858367 - -
          Affymetrix Rat Gene 1.0 ST Array 10858364 - -
          Affymetrix Rat Gene 1.0 ST Array 10702442 - -
          Affymetrix Rat Gene 1.0 ST Array 10858363 - -
          Affymetrix Rat Gene 1.0 ST Array 10858368 - -
          Affymetrix Rat Gene 1.0 ST Array 10858365 - -
          Affymetrix Rat Gene 1.0 ST Array 10858366 - -
          Affymetrix Rat Gene 1.1 ST Array 10858362 - -
          Affymetrix Rat Gene 1.1 ST Array 10702441 - -
          Rat Genome 230 2.0 Array 1372279_at - -
          Rat Genome 230 2.0 Array 1372279_PM_at - -
          Affymetrix HT Rat Focus Array 1372279_PM_at - -
          Affymetrix HT Rat Focus Array 1372279_at - -
          Affymetrix Rat Clariom S Assay PSR0400012119.rn.1 TC0400001695.rn.1 RTA TC
          Rat Expression Set 230 1372279_PM_at - -
          Rat Expression Set 230 1372279_at - -
          Affymetrix Rat Gene 1.0 ST Array 10858362 - -
          Rat Genome U34 Set rc_AA892271_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21435551 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369596 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline