Search
Search

Plekha5| Gene Symbol: Plekha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763357.1 | XP_008761579.1 | 24 - 25 | 3070 | 67 | |
| XM_008763358.1 | XP_008761580.1 | 25 - 26 | 3106 | 67 | ||
| XM_008763359.1 | XP_008761581.1 | 22 - 23 | 2869 | 67 | ||
| XM_008763360.1 | XP_008761582.1 | 21 - 22 | 2815 | 67 | ||
| XM_008763361.1 | XP_008761583.1 | 23 - 24 | 3797 | 67 | ||
| XM_008763362.1 | XP_008761584.1 | 19 - 20 | 2608 | 67 | ||
| XM_008763363.1 | XP_008761585.1 | 18 - 19 | 2934 | 67 | ||
| GenBank mRNA | BC158707.1 | - | 1 - 2 | 47 | 67 | |
Set Membership: |
|