Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01493430_g1
See other HEY1 GE Assays ›
Gene Symbol:
Hey1
Gene Name:
hes-related family bHLH transcription factor with YRPW motif 1
Gene Aliases:
Herp2
Chromosome Location:
Chr.2: 95320283 - 95322701 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01493430_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Hey1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001191845.1
NP_001178774.1
2 - 3
222
63
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0200000803.rn.1
Set Membership:
Amplicon length less than or equal to 70
Detect Genomic DNA
Rattus norvegicus
Probe spans exons
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
blood vessel development
vasculogenesis
osteoblast development
outflow tract morphogenesis
atrioventricular valve morphogenesis
pulmonary valve morphogenesis
atrioventricular valve formation
epithelial to mesenchymal transition involved in endocardial cushion formation
endocardial cushion to mesenchymal transition involved in heart valve formation
endocardial cushion morphogenesis
cardiac ventricle morphogenesis
ventricular trabecula myocardium morphogenesis
transcription, DNA-templated
transcription from RNA polymerase II promoter
Notch signaling pathway
anterior/posterior axis specification
negative regulation of gene expression
mesenchymal cell development
dorsal aorta morphogenesis
umbilical cord morphogenesis
negative regulation of neuron differentiation
positive regulation of osteoblast differentiation
negative regulation of Notch signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cardiac epithelial to mesenchymal transition
cardiac septum morphogenesis
ventricular septum morphogenesis
labyrinthine layer blood vessel development
artery development
arterial endothelial cell differentiation
cellular response to glucocorticoid stimulus
negative regulation of transcription regulatory region DNA binding
negative regulation of transcription from RNA polymerase II promoter involved in smooth muscle cell differentiation
regulation of vasculogenesis
transcription factor activity, RNA polymerase II core promoter sequence-specific
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
molecular_function
transcription factor activity, sequence-specific DNA binding
microsatellite binding
sequence-specific DNA binding
protein dimerization activity
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR0200004936.rn.1
TC0200000803.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline