Search
Search
| Gene Symbol: Lrrc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044696.1 | NP_001038161.1 | 6 - 7 | 783 | 133 | |
| XM_006232235.2 | XP_006232297.1 | 7 - 8 | 754 | 133 | ||
| XM_008760937.1 | XP_008759159.1 | 7 - 8 | 845 | 133 | ||
| XM_008760938.1 | XP_008759160.1 | 6 - 7 | 541 | 133 | ||
| XM_008760939.1 | XP_008759161.1 | 6 - 7 | 737 | 133 | ||
| GenBank mRNA | BC097432.1 | - | 6 - 7 | 783 | 7455342 | 133 |
Set Membership: |
|