Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01494892_m1
          See other TM4SF1 GE Assays ›
          Gene Symbol:
          Tm4sf1
          Gene Name:
          transmembrane 4 L six family member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 147382866 - 147392189 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01494892_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tm4sf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106434.1 NP_001099904.1 3 - 4 749 76
          XM_008760989.1 XP_008759211.1 3 - 4 876 76
          GenBank mRNA FQ220762.1 - 3 - 4 515 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003724.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58048 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11152 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11152 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11152 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11152 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41225 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823282 - -
          Affymetrix Rat Gene 1.0 ST Array 10823280 - -
          Affymetrix Rat Gene 1.0 ST Array 10823281 - -
          Affymetrix Rat Gene 1.0 ST Array 10823279 - -
          Affymetrix Rat Gene 1.0 ST Array 10823283 - -
          Affymetrix Rat Gene 1.1 ST Array 10823278 - -
          Rat Genome 230 2.0 Array 1373847_PM_at - -
          Rat Genome 230 2.0 Array 1378305_at - -
          Rat Genome 230 2.0 Array 1378305_PM_at - -
          Rat Genome 230 2.0 Array 1373847_at - -
          Affymetrix HT Rat Focus Array 1378305_at - -
          Affymetrix HT Rat Focus Array 1373847_PM_at - -
          Affymetrix HT Rat Focus Array 1373847_at - -
          Affymetrix HT Rat Focus Array 1378305_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200025004.rn.1 TC0200003724.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200025003.rn.1 TC0200003724.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200025005.rn.1 TC0200003724.rn.1 RTA TC
          Rat Expression Set 230 1373847_PM_at - -
          Rat Expression Set 230 1373847_at - -
          Rat Expression Set 230 1378305_PM_at - -
          Rat Expression Set 230 1378305_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823278 - -
          Rat Genome U34 Set rc_AI233280_at - -
          Rat Genome U34 Set rc_AI176174_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22422760 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361672 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline